Gene Transcripts

Gene Symbol
LIN52
Entrez Gene ID
91750
Gene Name
lin-52 DREAM MuvB core complex component
Gene Aliases
C14orf46, c14_5549
Chromosome Location
Chr. 14: 74084831 - 74201255 on Build GRCh38
UniGene ID
Hs.612866
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011537320.2 XP_011535622.1 6 4330


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CR616131.1 - 1 991 Chr. 14 - 74116918 - 74118235 [+]

View Full Product Details