Gene Transcripts
- Gene Symbol
- LINC01553
- Entrez Gene ID
- 283025
- Gene Name
- long intergenic non-protein coding RNA 1553
- Gene Aliases
- C10orf40
- Chromosome Location
- Chr. 10: 59958217 - 59960913 on Build GRCh38
- UniGene ID
- Hs.522955
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CR933684.1 |
- |
3 |
3105 |
|
View Full Product Details