Gene Transcripts

Gene Symbol
PHTF1
Entrez Gene ID
10745
Gene Name
putative homeodomain transcription factor 1
Gene Aliases
PHTF
Chromosome Location
Chr. 1: 113697202 - 113759888 on Build GRCh38
UniGene ID
Hs.655824
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK126773.1 - 1 1290


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK094327.1 - 1 890 Chr. 20 - 34796029 - 34798162 [-]

View Full Product Details