Gene Transcripts
- Gene Symbol
- BTBD19
- Entrez Gene ID
- 149478
- Gene Name
- BTB domain containing 19
- Gene Aliases
- Chromosome Location
- Chr. 1: 44807442 - 44814199 on Build GRCh38
- UniGene ID
- Hs.632400
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK056150.1 |
- |
4 |
2365 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BF828217.1 |
- |
1 |
458 |
|
Chr. 1 - 44814889 - 44815295 [+] |
GenBankmRNA |
BF828171.1 |
- |
1 |
482 |
|
Chr. 1 - 44814889 - 44815406 [+] |
GenBankmRNA |
BF828503.1 |
- |
1 |
458 |
|
Chr. 1 - 44814889 - 44815409 [+] |
GenBankmRNA |
BX105060.1 |
- |
1 |
252 |
|
Chr. 1 - 44815021 - 44815582 [+] |
View Full Product Details