Gene Transcripts
- Gene Symbol
- DCUN1D1
- Entrez Gene ID
- 54165
- Gene Name
- defective in cullin neddylation 1 domain containing 1
- Gene Aliases
- DCNL1, DCUN1L1, RP42, SCCRO, SCRO, Tes3
- Chromosome Location
- Chr. 3: 182938158 - 182986364 on Build GRCh38
- UniGene ID
- Hs.592789
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY994955.1 |
- |
1 |
480 |
|
Chr. 3 - 182938666 - 182939353 [-] |
GenBankmRNA |
BM717917.1 |
- |
1 |
355 |
|
Chr. 3 - 182938605 - 182939244 [-] |
GenBankmRNA |
AW900701.1 |
- |
1 |
200 |
|
Chr. 3 - 182938675 - 182939089 [-] |
View Full Product Details