Gene Transcripts
- Gene Symbol
- MED27
- Entrez Gene ID
- 9442
- Gene Name
- mediator complex subunit 27
- Gene Aliases
- CRAP34, CRSP34, CRSP8, MED3, TRAP37
- Chromosome Location
- Chr. 9: 131860110 - 132079887 on Build GRCh38
- UniGene ID
- Hs.374262
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK022289.1 |
- |
1 |
184 |
|
AK298436.1 |
- |
6 |
804 |
|
AK316345.1 |
- |
6 |
805 |
|
View Full Product Details