Gene Transcripts

Gene Symbol
DESI2
Entrez Gene ID
51029
Gene Name
desumoylating isopeptidase 2
Gene Aliases
C1orf121, CGI-146, DESI, DESI1, DeSI-2, FAM152A, PNAS-4, PPPDE1
Chromosome Location
Chr. 1: 244653048 - 244709034 on Build GRCh38
UniGene ID
Hs.498317
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011544203.2 XP_011542505.1 2 307
XM_011544206.2 XP_011542508.1 2 45
GenBank mRNA BC020640.1 - 1 814

View Full Product Details