Gene Transcripts
- Gene Symbol
- TSRM
- Entrez Gene ID
- 100287664
- Gene Name
- zinc finger domain-related protein TSRM
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.544905
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY742823.1 |
- |
1 |
103 |
|
View Full Product Details