Gene Transcripts

Gene Symbol
C7orf49
Entrez Gene ID
78996
Gene Name
chromosome 7 open reading frame 49
Gene Aliases
MRI
Chromosome Location
Chr. 7: 135092303 - 135170826 on Build GRCh38
UniGene ID
Hs.643113
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017012589.1 XP_016868078.1 4 5100
XM_017012590.1 XP_016868079.1 4 5026
XM_017012591.1 XP_016868080.1 4 4880
XM_017012592.1 XP_016868081.1 3 5061
XM_017012593.1 XP_016868082.1 4 4930
XM_017012594.1 XP_016868083.1 4 5137


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BU175034.1 - 1 87 Chr. 7 - 135161764 - 135162508 [-]
GenBankmRNA DA571644.1 - 1 65 Chr. 7 - 135161786 - 135162485 [-]
GenBankmRNA BU169260.1 - 1 96 Chr. 7 - 135161643 - 135162509 [-]
GenBankmRNA BG680106.1 - 1 40 Chr. 7 - 135161779 - 135162460 [-]
GenBankmRNA BQ672721.1 - 86 Chr. - - [-]
GenBankmRNA AK056910.1 - 1 1255 Chr. 7 - 135161548 - 135163675 [-]
GenBankmRNA DA571246.1 - 1 65 Chr. 7 - 135161607 - 135162485 [-]
GenBankmRNA BQ675467.1 - 87 Chr. - - [-]

View Full Product Details