Gene Transcripts
- Gene Symbol
- SNX24
- Entrez Gene ID
- 28966
- Gene Name
- sorting nexin 24
- Gene Aliases
- PRO1284, SBBI31
- Chromosome Location
- Chr. 5: 122845465 - 123029318 on Build GRCh38
- UniGene ID
- Hs.483200
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021473.1 |
- |
1 |
786 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB111567.1 |
- |
1 |
330 |
|
Chr. 5 - 123007059 - 123007618 [+] |
View Full Product Details