Gene Transcripts
- Gene Symbol
- PITPNC1
- Entrez Gene ID
- 26207
- Gene Name
- phosphatidylinositol transfer protein, cytoplasmic 1
- Gene Aliases
- M-RDGB-beta, MRDGBbeta, RDGB-BETA, RDGBB, RDGBB1
- Chromosome Location
- Chr. 17: 67377281 - 67697263 on Build GRCh38
- UniGene ID
- Hs.591185
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM690777.1 |
- |
1 |
606 |
|
Chr. 17 - 67696503 - 67697116 [+] |
View Full Product Details