Gene Transcripts

Gene Symbol
ARMCX4
Entrez Gene ID
100131755
Gene Name
armadillo repeat containing, X-linked 4
Gene Aliases
CXorf35, GASP4
Chromosome Location
Chr. X: 101418263 - 101535988 on Build GRCh38
UniGene ID
Hs.729062
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028407.2 16 6554
NR_045861.1 13 5140
NR_045862.1 13 4503
NR_045864.1 13 5182
GenBank mRNA AL050285.1 - 1 1934

View Full Product Details