Gene Transcripts

Gene Symbol
C14orf37
Entrez Gene ID
145407
Gene Name
chromosome 14 open reading frame 37
Gene Aliases
c14_5376
Chromosome Location
Chr. 14: 57993536 - 58152305 on Build GRCh38
UniGene ID
Hs.26766
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017021029.1 XP_016876518.1 8 12641
XM_017021030.1 XP_016876519.1 8 12781
GenBank mRNA BC031314.1 - 1 34

View Full Product Details