Gene Transcripts
- Gene Symbol
- CSNK1A1
- Entrez Gene ID
- 1452
- Gene Name
- casein kinase 1 alpha 1
- Gene Aliases
- CK1, CK1a, CKIa, HEL-S-77p, HLCDGP1, PRO2975
- Chromosome Location
- Chr. 5: 149495894 - 149551552 on Build GRCh38
- UniGene ID
- Hs.529862
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000499521.2 |
- |
2 |
3940 |
|
Chr. 5 - 149494314 - 149504670 [-] |
GenBankmRNA |
L26953.1 |
- |
|
397 |
|
Chr. - - [-] |
GenBankmRNA |
AL699470.1 |
- |
1 |
352 |
|
Chr. 5 - 149498797 - 149499361 [-] |
View Full Product Details