Gene Transcripts
- Gene Symbol
- MTHFD2L
- Entrez Gene ID
- 441024
- Gene Name
- methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like
- Gene Aliases
- Chromosome Location
- Chr. 4: 74123280 - 74303099 on Build GRCh38
- UniGene ID
- Hs.721011
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021544.1 |
- |
1 |
1854 |
|
BC037529.2 |
- |
7 |
1001 |
|
View Full Product Details