Gene Transcripts
- Gene Symbol
- Tnrc6b
- Entrez Gene ID
- 213988
- Gene Name
- trinucleotide repeat containing 6b
- Gene Aliases
- 2700090M07Rik, A730065C02Rik, AI848765, D230019K20Rik
- Chromosome Location
- Chr. 15: 80711313 - 80941086 on Build GRCm38
- UniGene ID
- Mm.131328
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK012593.1 |
- |
1 |
282 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY711738.1 |
- |
1 |
279 |
|
Chr. 15 - 80884353 - 80884727 [+] |
View Full Product Details