Gene Transcripts
- Gene Symbol
- Trio
- Entrez Gene ID
- 223435
- Gene Name
- triple functional domain (PTPRF interacting)
- Gene Aliases
- 6720464I07Rik, AA408740, Solo
- Chromosome Location
- Chr. 15: 27730649 - 28025953 on Build GRCm38
- UniGene ID
- Mm.485422
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020139.1 |
- |
1 |
891 |
|
View Full Product Details