Gene Transcripts
- Gene Symbol
- Pgls
- Entrez Gene ID
- 66171
- Gene Name
- 6-phosphogluconolactonase
- Gene Aliases
- 1110030K05Rik, AI447866, Plgs
- Chromosome Location
- Chr. 8: 71592184 - 71596267 on Build GRCm38
- UniGene ID
- Mm.282284
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK080749.1 |
- |
|
956 |
|
View Full Product Details