Gene Transcripts

Gene Symbol
Thap8
Entrez Gene ID
75629
Gene Name
THAP domain containing 8
Gene Aliases
1700020L13Rik
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.271677
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK006173.1 - 2, 3 479


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY706272.1 - 2, 3 479 Chr. 7 - 30280094 - 30289704 [+]

View Full Product Details