Gene Transcripts
- Gene Symbol
- Thap8
- Entrez Gene ID
- 75629
- Gene Name
- THAP domain containing 8
- Gene Aliases
- 1700020L13Rik
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.271677
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK006173.1 |
- |
1 |
267 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY706272.1 |
- |
1 |
267 |
|
Chr. 7 - 30280094 - 30289704 [+] |
View Full Product Details