Gene Transcripts
- Gene Symbol
- Mcu
- Entrez Gene ID
- 215999
- Gene Name
- mitochondrial calcium uniporter
- Gene Aliases
- 2010012O16Rik, AV064928, C10orf42, Ccdc109a, D130073L02Rik, Gm64
- Chromosome Location
- Chr. 10: 59446984 - 59616692 on Build GRCm38
- UniGene ID
- Mm.296470
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK166044.1 |
- |
1, 2 |
161 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY734336.1 |
- |
1, 2 |
161 |
|
Chr. 10 - 59534799 - 59616675 [-] |
View Full Product Details