Gene Transcripts
- Gene Symbol
- Tbx1
- Entrez Gene ID
- 21380
- Gene Name
- T-box 1
- Gene Aliases
- Chromosome Location
- Chr. 16: 18581704 - 18590671 on Build GRCm38
- UniGene ID
- Mm.295194
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK029096.1 |
- |
|
1845 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB009722.2 |
- |
1 |
132 |
|
Chr. 16 - 18580980 - 18581615 [-] |
View Full Product Details