Gene Transcripts
- Gene Symbol
- Sike1
- Entrez Gene ID
- 66641
- Gene Name
- suppressor of IKBKE 1
- Gene Aliases
- 2810005O12Rik, 5730470L24Rik, AI450236, AI839862, Sike, Sikeb
- Chromosome Location
- Chr. 3: 102995740 - 103003914 on Build GRCm38
- UniGene ID
- Mm.208619
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK139055.1 |
- |
|
1172 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CF551415.1 |
- |
1 |
143 |
|
Chr. 3 - 103007165 - 103008013 [+] |
View Full Product Details