Gene Transcripts
- Gene Symbol
- Srcap
- Entrez Gene ID
- 100043597
- Gene Name
- Snf2-related CREBBP activator protein
- Gene Aliases
- B930091H02Rik, D030022P06Rik, F630004O05Rik
- Chromosome Location
- Chr. 7: 127511983 - 127561219 on Build GRCm38
- UniGene ID
- Mm.428042
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK081131.1 |
- |
|
727 |
|
AK086186.1 |
- |
|
717 |
|
View Full Product Details