Gene Transcripts
- Gene Symbol
- Maml2
- Entrez Gene ID
- 270118
- Gene Name
- mastermind like 2 (Drosophila)
- Gene Aliases
- 5930431H10, BC032967
- Chromosome Location
- Chr. 9: 13297958 - 13298561 on Build GRCm38
- UniGene ID
- Mm.116898
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK040022.1 |
- |
|
433 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB634845.1 |
- |
2 |
433 |
|
Chr. 9 - 13298193 - 13520103 [+] |
GenBankmRNA |
BB202315.2 |
- |
2 |
444 |
|
Chr. 9 - 13298193 - 13520101 [+] |
View Full Product Details