Gene Transcripts
- Gene Symbol
- PPIEL
- Entrez Gene ID
- 728448
- Gene Name
- peptidylprolyl isomerase E like pseudogene
- Gene Aliases
- PPIEP1
- Chromosome Location
- Chr. 1: 39522280 - 39559698 on Build GRCh38
- UniGene ID
- Hs.472508
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK093659.1 |
- |
1 |
611 |
|
View Full Product Details