Gene Transcripts
- Gene Symbol
- ATP1A1-AS1
- Entrez Gene ID
- 84852
- Gene Name
- ATP1A1 antisense RNA 1
- Gene Aliases
- ATP1A1OS, C1orf203
- Chromosome Location
- Chr. 1: 116392865 - 116418622 on Build GRCh38
- UniGene ID
- Hs.664752
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK309389.1 |
- |
3 |
948 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000608511.5 |
- |
3 |
948 |
|
Chr. 1 - 116392247 - 116418622 [-] |
View Full Product Details