Gene Transcripts

Gene Symbol
CDC14A
Entrez Gene ID
8556
Gene Name
cell division cycle 14A
Gene Aliases
DFNB105, cdc14, hCDC14
Chromosome Location
Chr. 1: 100345015 - 100520281 on Build GRCh38
UniGene ID
Hs.127411
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319211.1 NP_001306140.1 1 149
GenBank mRNA AK302310.1 - 1 138
AK310292.1 - 1 126
DB029799.1 - 1 162


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000446055.1 - 2 388 Chr. 1 - 100344477 - 100345242 [+]

View Full Product Details