Gene Transcripts

Gene Symbol
HAR1A
Entrez Gene ID
768096
Gene Name
highly accelerated region 1A (non-protein coding)
Gene Aliases
HAR1F, LINC00064, NCRNA00064
Chromosome Location
Chr. 20: 63101292 - 63104385 on Build GRCh38
UniGene ID
Hs.549606
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003244.1
2 2767
GenBank mRNA BC047717.1 -
2 1847
DQ860409.1 - LIT3337
HAR1F
2 2767


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000433161.1 - 2 1854 Chr. 20 - 63102205 - 63104386 [+]

View Full Product Details