Gene Transcripts
- Gene Symbol
- RPSAP41
- Entrez Gene ID
- 100270918
- Gene Name
- ribosomal protein SA pseudogene 41
- Gene Aliases
- RPSA_17_711
- Chromosome Location
- Chr. 6: 73290550 - 73291335 on Build GRCh38
- UniGene ID
- Hs.661304
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL832252.1 |
- |
1 |
2381 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000421315.1 |
- |
1 |
2379 |
|
Chr. 6 - 73268051 - 73291340 [+] |
GENCODE |
ENST00000442007.1 |
- |
2 |
234 |
|
Chr. 6 - 73263215 - 73276517 [+] |
View Full Product Details