Gene Transcripts
- Gene Symbol
- GS1-600G8.3
- Entrez Gene ID
- 100093698
- Gene Name
- unknown transcript
- Gene Aliases
- Chromosome Location
- Chr. X: 13310652 - 13319933 on Build GRCh38
- UniGene ID
- Hs.643612
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046087.1 |
|
15 |
1419 |
|
GenBank mRNA |
BC022040.1 |
- |
15 |
1424 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000431486.1 |
- |
15 |
1419 |
|
Chr. X - 13310652 - 13319933 [+] |
View Full Product Details