Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_024340.1 | 2 | 1637 | ||
GenBank mRNA | BC038741.1 | - | 2 | 1638 | |
BC048976.2 | - | 1 | 1576 | ||
BC119749.2 | - | 1 | 513 | ||
BC119750.2 | - | 1 | 513 | ||
BC126326.1 | - | 1 | 848 | ||
BC130402.1 | - | 1 | 848 | ||
CR933684.1 | - | 3 | 1363 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000444900.1 | - | 2 | 1637 | Chr. 10 - 59958217 - 59960913 [-] |