Gene Transcripts
- Gene Symbol
- DNM1P35
- Entrez Gene ID
- 100128285
- Gene Name
- dynamin 1 pseudogene 35
- Gene Aliases
- DNM1DN8-2, DNM1DN8.2, DNM1DN8@, FKSG88
- Chromosome Location
- Chr. 15: 75727670 - 75740077 on Build GRCh38
- UniGene ID
- Hs.604066
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024595.3 |
|
5 |
876 |
|
GenBank mRNA |
AK091244.1 |
- |
3 |
497 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000501931.1 |
- |
3 |
497 |
|
Chr. 15 - 75727670 - 75738623 [-] |
View Full Product Details