Gene Transcripts
- Gene Symbol
- SRP14-AS1
- Entrez Gene ID
- 100131089
- Gene Name
- SRP14 antisense RNA1 (head to head)
- Gene Aliases
- Chromosome Location
- Chr. 15: 40039311 - 40067509 on Build GRCh38
- UniGene ID
- Hs.386000
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040059.1 |
|
3 |
441 |
|
GenBank mRNA |
EB388563.1 |
- |
3 |
365 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000559012.1 |
- |
1 |
126 |
|
Chr. 15 - 40064387 - 40065705 [+] |
View Full Product Details