Gene Transcripts
- Gene Symbol
- GS1-120K12.4
- Entrez Gene ID
- 101929069
- Gene Name
- uncharacterized GS1-120K12.4
- Gene Aliases
- Chromosome Location
- Chr. 1: 185001527 - 185008678 on Build GRCh38
- UniGene ID
- Hs.497170
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF508903.1 |
- |
2 |
543 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000566476.1 |
- |
2 |
543 |
|
Chr. 1 - 185001527 - 185008683 [+] |
View Full Product Details