Gene Transcripts
- Gene Symbol
- Ptf1aos
- Entrez Gene ID
- 102637107
- Gene Name
- pancreas specific transcription factor 1a, opposite strand
- Gene Aliases
- Gm13336, OTTMUSG00000011493
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.406835
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB701680.1 |
- |
4 |
891 |
|
AK053418.1 |
- |
4 |
891 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB539064.2 |
- |
4 |
612 |
|
Chr. 2 - 19441508 - 19446396 [-] |
View Full Product Details