Gene Transcripts

Gene Symbol
Prss29
Entrez Gene ID
114662
Gene Name
protease, serine 29
Gene Aliases
D19363, Isp2, mISP-2
Chromosome Location
Chr. 17: 25318654 - 25322684 on Build GRCm38
UniGene ID
Mm.207082
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053260.3 NP_444490.2 4 576
GenBank mRNA AF305425.2 - 3 412
AF442819.1 - 3 412
AK144368.1 - 4 577
BC104122.1 - 3 412 40044207
BC104123.1 - 3 412

View Full Product Details