Gene Transcripts

Gene Symbol
Dcun1d1
Entrez Gene ID
114893
Gene Name
DCN1, defective in cullin neddylation 1, domain containing 1 (S. cerevisiae)
Gene Aliases
Rp42, SCCRO, Tes3, pTes3
Chromosome Location
Chr. 3: 35892103 - 35937605 on Build GRCm38
UniGene ID
Mm.374810
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205361.1 NP_001192290.1 3, 4 569
NM_001205362.1 NP_001192291.1 3, 4 529
NM_033623.6 NP_296372.3 3, 4 516
GenBank mRNA AF198092.1 - 3, 4 407
AJ305034.1 - 3, 4 569
AK008657.1 - 3, 4 406
AK139845.1 - 3, 4 519
AK155081.1 - 3, 4 440
AK165651.1 - 3, 4 442
AK165675.1 - 3, 4 414
BC020161.1 - 3, 4 492 3988680
BC031666.1 - 3, 4 425 4012394
BC037431.1 - 3, 4 425 2648141

Gene Symbol
Tes3-ps
Entrez Gene ID
54339
Gene Name
testis derived transcript 3, pseudogene
Gene Aliases
Chromosome Location
Chr. 13: 49493652 - 49496389 on Build GRCm38
UniGene ID
Mm.441473
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK029314.1 - 1 722

View Full Product Details