Gene Transcripts
- Gene Symbol
- Vmn1r27
- Entrez Gene ID
- 171206
- Gene Name
- vomeronasal 1 receptor 27
- Gene Aliases
- V1rc33
- Chromosome Location
- Chr. 6: 58215106 - 58216017 on Build GRCm38
- UniGene ID
- Mm.389781
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134436.2 |
NP_602318.2
|
1 |
367 |
|
- Gene Symbol
- Vmn1r20
- Entrez Gene ID
- 434017
- Gene Name
- vomeronasal 1 receptor 20
- Gene Aliases
- EG434017, Gm5569
- Chromosome Location
- Chr. 6: 57431691 - 57432602 on Build GRCm38
- UniGene ID
- Mm.473685
- Species
- Mus musculus
View Full Product Details