Gene Transcripts

Gene Symbol
Vmn1r3
Entrez Gene ID
100312471
Gene Name
vomeronasal 1 receptor 3
Gene Aliases
Gm11778, OTTMUSG00000004181
Chromosome Location
Chr. 4: 3184385 - 3185305 on Build GRCm38
UniGene ID
Mm.377218
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167535.1 NP_001161007.1 1 489
GenBank mRNA BC145887.1 - 1 542

Gene Symbol
Vmn1r2
Entrez Gene ID
100312470
Gene Name
vomeronasal 1 receptor 2
Gene Aliases
Gm11776, OTTMUSG00000004179
Chromosome Location
Chr. 4: 3172083 - 3173003 on Build GRCm38
UniGene ID
Mm.480297
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167534.1 NP_001161006.1 1 489

Gene Symbol
Vmn1r238
Entrez Gene ID
100312476
Gene Name
vomeronasal 1 receptor, 238
Gene Aliases
Chromosome Location
Chr. 18: 3122492 - 3123412 on Build GRCm38
UniGene ID
Mm.480293
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167539.1 NP_001161011.1 1 489

Gene Symbol
Vmn1r79
Entrez Gene ID
100042437
Gene Name
vomeronasal 1 receptor 79
Gene Aliases
Gm9807
Chromosome Location
Chr. 7: 12176193 - 12177113 on Build GRCm38
UniGene ID
Mm.484984
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166835.1 NP_001160307.1 1 489

View Full Product Details