Gene Transcripts
- Gene Symbol
- Pkd1l1
- Entrez Gene ID
- 171395
- Gene Name
- polycystic kidney disease 1 like 1
- Gene Aliases
- AA444596
- Chromosome Location
- Chr. 11: 8813839 - 8974367 on Build GRCm38
- UniGene ID
- Mm.146711
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK140049.1 |
- |
10 |
1167 |
|
View Full Product Details