Gene Transcripts

Gene Symbol
Tet3
Entrez Gene ID
194388
Gene Name
tet methylcytosine dioxygenase 3
Gene Aliases
B430006D22Rik, BC037432, D230004J03Rik
Chromosome Location
Chr. 6: 83362373 - 83457313 on Build GRCm38
UniGene ID
Mm.211030
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183138.2 NP_898961.2 4, 5 2435
GenBank mRNA HQ423151.1 - 4, 5 2609
HQ423152.1 - 3, 4 2552
JX946278.1 - 5, 6 3141

View Full Product Details