Gene Transcripts
- Gene Symbol
- PDHB
- Entrez Gene ID
- 5162
- Gene Name
- pyruvate dehydrogenase (lipoamide) beta
- Gene Aliases
- PDHBD, PDHE1-B, PHE1B
- Chromosome Location
- Chr. 3: 58427630 - 58433852 on Build GRCh38
- UniGene ID
- Hs.161357
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000925.3 |
NP_000916.2
|
2, 3 |
150 |
|
NM_001173468.1 |
NP_001166939.1
|
2, 3 |
150 |
|
NM_001315536.1 |
NP_001302465.1
|
1, 2 |
96 |
|
NR_033384.1 |
|
1, 2 |
220 |
|
GenBank mRNA |
AB209241.1 |
- |
2 |
1760 |
|
AK225621.1 |
- |
2, 3 |
130 |
|
AK293153.1 |
- |
2, 3 |
116 |
|
AK311514.1 |
- |
2, 3 |
125 |
|
AK313022.1 |
- |
2, 3 |
125 |
|
AL117618.1 |
- |
1, 2 |
61 |
|
BC000439.2 |
- |
2, 3 |
117 |
2820672 |
BC001924.1 |
- |
2, 3 |
117 |
2820672 |
CR541911.1 |
- |
2, 3 |
107 |
|
DA965753.1 |
- |
2, 3 |
150 |
|
DB039833.1 |
- |
1, 2 |
220 |
|
J03576.1 |
- |
2, 3 |
125 |
|
M34055.1 |
- |
2, 3 |
112 |
|
M34056.1 |
- |
1, 2 |
35 |
|
M34479.1 |
- |
2, 3 |
107 |
|
M54788.1 |
- |
2, 3 |
112 |
|
X57778.1 |
- |
2, 3 |
126 |
|
View Full Product Details