Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195265.1 | NP_001182194.1 | 16 | 3052 | |
GenBank mRNA | AK033653.1 | - | 6 | 947 | |
BC096488.1 | - | 6 | 1168 | ||
BC147398.1 | - | 3 | 855 | 9056452 | |
BC147399.1 | - | 3 | 855 | 9056443 | |
BC172054.1 | - | 3 | 855 | ||
KF041446.1 | - | 17 | 3018 | ||
KF041447.1 | - | 17 | 3047 | ||
KF041448.1 | - | 14 | 2832 | ||
KF041449.1 | - | 15 | 2865 | ||
KF041450.1 | - | 16 | 2957 | ||
KF041451.1 | - | 17 | 2838 |