Gene Transcripts

Gene Symbol
Usp43
Entrez Gene ID
216835
Gene Name
ubiquitin specific peptidase 43
Gene Aliases
Chromosome Location
Chr. 11: 67854523 - 67922153 on Build GRCm38
UniGene ID
Mm.158885
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291049.1 NP_001277978.1 15 2710
NM_173754.4 NP_776115.2 15 2725
GenBank mRNA AK042217.1 - 15 2711
AK047339.1 - 3 697
AK083271.1 - 15 2563
BC021474.1 - 14 2015 5343429

View Full Product Details