Gene Transcripts

Gene Symbol
Unc79
Entrez Gene ID
217843
Gene Name
unc-79 homolog (C. elegans)
Gene Aliases
9030205A07Rik, Mlca3
Chromosome Location
Chr. 12: 102948329 - 103184228 on Build GRCm38
UniGene ID
Mm.387073
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081017.2 NP_001074486.2 15 2529
GenBank mRNA AB257853.1 - 15 2529
AK144534.1 - 4 952
GQ334471.1 - 15 1675

View Full Product Details