Gene Transcripts

Gene Symbol
Trio
Entrez Gene ID
223435
Gene Name
triple functional domain (PTPRF interacting)
Gene Aliases
6720464I07Rik, AA408740, Solo
Chromosome Location
Chr. 15: 27730649 - 28025953 on Build GRCm38
UniGene ID
Mm.485422
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081302.1 NP_001074771.1 38, 39 6269
GenBank mRNA BC060724.1 - 18, 19 2485

View Full Product Details