Gene Transcripts

Gene Symbol
Zc3h3
Entrez Gene ID
223642
Gene Name
zinc finger CCCH type containing 3
Gene Aliases
BC049953, Kiaa0150, Smicl, Zc3hdc3
Chromosome Location
Chr. 15: 75754434 - 75841971 on Build GRCm38
UniGene ID
Mm.209800
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172121.1 NP_742119.1 2 923
GenBank mRNA AJ516034.1 - 2 923
AK138281.1 - 2 931
BC060682.1 - 2 922 6416246

View Full Product Details