Gene Transcripts

Gene Symbol
Nmnat2
Entrez Gene ID
226518
Gene Name
nicotinamide nucleotide adenylyltransferase 2
Gene Aliases
AI843915, D030041I09Rik, PNAT1, PNAT2
Chromosome Location
Chr. 1: 152954940 - 153119266 on Build GRCm38
UniGene ID
Mm.40548
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175460.3 NP_780669.1 1 63
GenBank mRNA AK083532.1 - 1 167
AK084041.1 - 1 173
AK129155.1 - 1 149
BC089007.1 - 1 63 30532596

View Full Product Details