Gene Transcripts
- Gene Symbol
- Npepl1
- Entrez Gene ID
- 228961
- Gene Name
- aminopeptidase-like 1
- Gene Aliases
- BC023239, C85514, mKIAA1974
- Chromosome Location
- Chr. 2: 174110335 - 174122702 on Build GRCm38
- UniGene ID
- Mm.295629
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_213733.2 |
NP_998898.1
|
4, 5 |
679 |
|
GenBank mRNA |
AK144651.1 |
- |
4, 5 |
652 |
|
AK150313.1 |
- |
4, 5 |
680 |
|
AK156136.1 |
- |
4, 5 |
680 |
|
AK170509.1 |
- |
4, 5 |
680 |
|
AK210473.1 |
- |
2, 3 |
184 |
|
BC023239.1 |
- |
3, 4 |
357 |
|
BC055448.1 |
- |
3, 4 |
354 |
|
BC069933.1 |
- |
4, 5 |
674 |
4008091 |
BC080707.1 |
- |
4, 5 |
552 |
|
View Full Product Details